Assay Details
| Gene Symbol: INTS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101108.1 | NP_001094578.1 | 10 - 11 | 1135 | 84 | |
| GenBank mRNA | BC110155.1 | - | 10 - 11 | 1146 | 84 | |
| BT030671.1 | - | 10 - 11 | 1135 | 84 | ||
More Information
Set Membership: |
|

INTS11
Genomic Map