Assay Details
| Gene Symbol: HEXD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101864.2 | NP_001095334.2 | 9 - 10 | 1741 | 87 | |
| GenBank mRNA | BC148958.1 | - | 9 - 10 | 1039 | 87 | |
More Information
Set Membership: |
|

HEXD
Genomic Map