Assay Details
| Gene Symbol: CCKAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101865.2 | NP_001095335.1 | 1 - 2 | 130 | 89 | |
| GenBank mRNA | BC147963.1 | - | 1 - 2 | 131 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC147963.1, NM_001101865.2 transcripts.
Set Membership: |
|

CCKAR
Genomic Map