Assay Details
| Gene Symbol: DDAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102201.2 | NP_001095671.1 | 5 - 6 | 835 | 65 | |
| GenBank mRNA | BC107532.1 | - | 5 - 6 | 832 | 65 | |
| BC149998.1 | - | 5 - 6 | 836 | 65 | ||
More Information
Set Membership: |
|

DDAH1
Genomic Map