Assay Details
| Gene Symbol: MAGED4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103311.1 | NP_001096781.1 | 10 - 11 | 1897 | 90 | |
| GenBank mRNA | BC147977.1 | - | 10 - 11 | 1897 | 90 | |
More Information
Set Membership: |
|

MAGED4B
Genomic Map