Assay Details
| Gene Symbol: ACTR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103338.1 | NP_001096808.1 | 2 - 3 | 216 | 71 | |
| GenBank mRNA | BC134541.1 | - | 2 - 3 | 216 | 71 | |
More Information
Set Membership: |
|

ACTR1B
Genomic Map