Assay Details
| Gene Symbol: ALG12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103346.1 | NP_001096816.1 | 3 - 4 | 518 | 98 | |
| GenBank mRNA | BC142132.1 | - | 3 - 4 | 518 | 98 | |
More Information
Set Membership: |
|

ALG12
Genomic Map