Assay Details
| Gene Symbol: TTC9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105507.1 | NP_001098977.1 | 2 - 3 | 616 | 94 | |
| GenBank mRNA | BC142207.1 | - | 2 - 3 | 616 | 94 | |
More Information
Set Membership: |
|

TTC9B
Genomic Map