Assay Details
| Gene Symbol: NUDT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105652.2 | NP_001099122.1 | 1 - 2 | 84 | 95 | |
| GenBank mRNA | BC122769.1 | - | 1 - 2 | 86 | 95 | |
More Information
Set Membership: |
|

NUDT14
Genomic Map