Assay Details
| Gene Symbol: ELANE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105653.1 | NP_001099123.1 | 1 - 2 | 88 | 114 | |
| GenBank mRNA | BC149420.1 | - | 1 - 2 | 88 | 114 | |
More Information
Set Membership: |
|

ELANE
Genomic Map