Assay Details
| Gene Symbol: ANAPC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113229.2 | NP_001106700.1 | 2 - 3 | 203 | 107 | |
| NM_001113234.2 | NP_001106705.1 | 2 - 3 | 246 | 107 | ||
| GenBank mRNA | BC111620.1 | - | 2 - 3 | 174 | 107 | |
| FE003831.1 | - | 2 - 3 | 200 | 107 | ||
More Information
Set Membership: |
|

ANAPC13
Genomic Map