Assay Details
| Gene Symbol: ATP5F1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143741.1 | NP_001137213.1 | 1 - 2 | 290 | 72 | |
| GenBank mRNA | BC108146.1 | - | 1 - 2 | 290 | 72 | |
| X16978.1 | - | 59 | 72 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC108146.1, NM_001143741.1 transcripts.
Set Membership: |
|

ATP5F1E
Genomic Map