Assay Details
| Gene Symbol: ATP5F1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143741.1 | NP_001137213.1 | 2 - 3 | 418 | 98 | |
| GenBank mRNA | BC108146.1 | - | 2 - 3 | 418 | 98 | |
| X16978.1 | - | 187 | 98 | |||
More Information
Set Membership: |
|

ATP5F1E
Genomic Map