Assay Details
| Gene Symbol: EIF1AY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145757.1 | NP_001139229.1 | 225 | 91 | ||
| GenBank mRNA | FJ627277.1 | - | 216 | 91 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EIF1AY| Gene Symbol: EIF1AY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145757.1 | NP_001139229.1 | 225 | 91 | ||
| GenBank mRNA | FJ627277.1 | - | 216 | 91 | ||
Set Membership: |
|