Assay Details
| Gene Symbol: TENT4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160157.1 | NP_001153629.1 | 3 - 4 | 259 | 58 | |
| GenBank mRNA | DT855254.1 | - | 2 - 3 | 161 | 58 | |
More Information
Set Membership: |
|

TENT4A
Genomic Map