Assay Details
| Gene Symbol: GPX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163142.2 | NP_001156614.1 | 4 - 5 | 476 | 85 | |
| GenBank mRNA | DY110071.1 | - | 4 - 5 | 471 | 85 | |
| DY121921.1 | - | 2 - 3 | 147 | 85 | ||
| DY143862.1 | - | 1 - 2 | 94 | 85 | ||
More Information
Set Membership: |
|

GPX6
Genomic Map