Assay Details
| Gene Symbol: C1H21orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163694.1 | NP_001157166.1 | 1 - 2 | 331 | 66 | |
| GenBank mRNA | AJ673870.1 | - | 1 - 2 | 331 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001163694.1 transcripts.
Set Membership: |
|

C1H21orf62
Genomic Map