Assay Details
| Gene Symbol: BEX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163777.2 | NP_001157249.1 | 1 - 2 | 74 | 62 | |
| GenBank mRNA | BC103258.1 | - | 1 - 2 | 75 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103258.1, NM_001163777.2 transcripts.
Set Membership: |
|

BEX3
Genomic Map