Assay Details
| Gene Symbol: TP53INP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205423.1 | NP_001192352.1 | 2 - 3 | 468 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TP53INP1| Gene Symbol: TP53INP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205423.1 | NP_001192352.1 | 2 - 3 | 468 | 69 | |
Set Membership: |
|