Assay Details
| Gene Symbol: IL17RD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205427.2 | NP_001192356.2 | 8 - 9 | 744 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IL17RD| Gene Symbol: IL17RD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205427.2 | NP_001192356.2 | 8 - 9 | 744 | 61 | |
Set Membership: |
|