Assay Details
| Gene Symbol: RMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205470.1 | NP_001192399.1 | 5 - 6 | 548 | 95 | |
| GenBank mRNA | BC151667.1 | - | 5 - 6 | 520 | 94 | |
More Information
Set Membership: |
|

RMC1
Genomic Map