Assay Details
| Gene Symbol: MAP3K20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205493.1 | NP_001192422.1 | 3 - 4 | 419 | 124 | |
| GenBank mRNA | BC126764.1 | - | 3 - 4 | 367 | 124 | |
More Information
Set Membership: |
|

MAP3K20
Genomic Map