Assay Details
| Gene Symbol: NCKAP5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205539.1 | NP_001192468.1 | 6 - 7 | 3092 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NCKAP5L| Gene Symbol: NCKAP5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205539.1 | NP_001192468.1 | 6 - 7 | 3092 | 68 | |
Set Membership: |
|