Assay Details
| Gene Symbol: ENTHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205542.1 | NP_001192471.1 | 4 - 5 | 831 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ENTHD1| Gene Symbol: ENTHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205542.1 | NP_001192471.1 | 4 - 5 | 831 | 100 | |
Set Membership: |
|