Assay Details
| Gene Symbol: SPAG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205549.1 | NP_001192478.1 | 4 - 5 | 1475 | 70 | |
| GenBank mRNA | BC149757.1 | - | 4 - 5 | 1481 | 70 | |
More Information
Set Membership: |
|

SPAG5
Genomic Map