Assay Details
| Gene Symbol: TRIM68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205571.1 | NP_001192500.1 | 5 - 6 | 928 | 92 | |
| GenBank mRNA | BC114155.1 | - | 5 - 6 | 927 | 92 | |
More Information
Set Membership: |
|

TRIM68
Genomic Map