Assay Details
| Gene Symbol: PCK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205594.1 | NP_001192523.1 | 7 - 8 | 1350 | 75 | |
| GenBank mRNA | BC102244.1 | - | 5 - 6 | 1936 | 75 | |
More Information
Set Membership: |
|

PCK2
Genomic Map