Assay Details
| Gene Symbol: BICDL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205599.1 | NP_001192528.1 | 7 - 8 | 1178 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BICDL2| Gene Symbol: BICDL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205599.1 | NP_001192528.1 | 7 - 8 | 1178 | 74 | |
Set Membership: |
|