Assay Details
| Gene Symbol: RETREG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205608.2 | NP_001192537.1 | 8 - 9 | 1065 | 76 | |
| GenBank mRNA | BT026241.1 | - | 3 - 4 | 371 | 76 | |
More Information
Set Membership: |
|

RETREG2
Genomic Map