Assay Details
| Gene Symbol: SLC39A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205648.1 | NP_001192577.1 | 3 - 4 | 630 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC39A2| Gene Symbol: SLC39A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205648.1 | NP_001192577.1 | 3 - 4 | 630 | 103 | |
Set Membership: |
|