Assay Details
| Gene Symbol: ADCK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205658.1 | NP_001192587.1 | 5 - 6 | 1550 | 62 | |
| GenBank mRNA | BC149001.1 | - | 6 - 7 | 2107 | 62 | |
More Information
Set Membership: |
|

ADCK2
Genomic Map