Assay Details
| Gene Symbol: TUT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205752.2 | NP_001192681.1 | 23 - 24 | 3968 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TUT7| Gene Symbol: TUT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205752.2 | NP_001192681.1 | 23 - 24 | 3968 | 94 | |
Set Membership: |
|