Assay Details
| Gene Symbol: TP53I13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205759.1 | NP_001192688.1 | 6 - 7 | 1224 | 77 | |
| GenBank mRNA | BC149397.1 | - | 6 - 7 | 1107 | 77 | |
More Information
Set Membership: |
|

TP53I13
Genomic Map