Assay Details
| Gene Symbol: FAM21A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205767.2 | NP_001192696.1 | 8 - 9 | 834 | 95 | |
| GenBank mRNA | BC151722.1 | - | 8 - 9 | 835 | 95 | |
More Information
Set Membership: |
|

FAM21A
Genomic Map