Assay Details
| Gene Symbol: KLHL22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205773.4 | NP_001192702.1 | 3 - 4 | 1154 | 69 | |
| GenBank mRNA | BT025359.1 | - | 4 - 5 | 1550 | 69 | |
More Information
Set Membership: |
|

KLHL22
Genomic Map