Assay Details
| Gene Symbol: TNFSF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205782.1 | NP_001192711.1 | 3 - 4 | 314 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNFSF15| Gene Symbol: TNFSF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205782.1 | NP_001192711.1 | 3 - 4 | 314 | 104 | |
Set Membership: |
|