Assay Details
| Gene Symbol: OSGEPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205798.1 | NP_001192727.1 | 2 - 3 | 360 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OSGEPL1| Gene Symbol: OSGEPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205798.1 | NP_001192727.1 | 2 - 3 | 360 | 73 | |
Set Membership: |
|