Assay Details
| Gene Symbol: MARCHF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205812.3 | NP_001192741.1 | 18 - 19 | 1703 | 63 | |
| GenBank mRNA | BC119893.1 | - | 8 - 9 | 776 | 63 | |
More Information
Set Membership: |
|

MARCHF6
Genomic Map