Assay Details
| Gene Symbol: ZNF445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205819.3 | NP_001192748.1 | 1 - 2 | 35 | 58 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001205819.3 transcripts.
Set Membership: |
|

ZNF445
Genomic Map