Assay Details
| Gene Symbol: ZNF445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205819.3 | NP_001192748.1 | 2 - 3 | 94 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001205819.3 transcripts.
Set Membership: |
|

ZNF445
Genomic Map