Assay Details
| Gene Symbol: PNLIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205820.1 | NP_001192749.1 | 12 - 13 | 1351 | 64 | |
| GenBank mRNA | HM751938.1 | - | 10 - 11 | 1283 | 64 | |
More Information
Set Membership: |
|

PNLIP
Genomic Map