Assay Details
| Gene Symbol: ANKRD33B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205847.3 | NP_001192776.3 | 2 - 3 | 495 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD33B| Gene Symbol: ANKRD33B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205847.3 | NP_001192776.3 | 2 - 3 | 495 | 84 | |
Set Membership: |
|