Assay Details
| Gene Symbol: VSIG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205873.1 | NP_001192802.1 | 4 - 5 | 790 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VSIG8| Gene Symbol: VSIG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205873.1 | NP_001192802.1 | 4 - 5 | 790 | 102 | |
Set Membership: |
|