Assay Details
| Gene Symbol: BOLA-DOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205920.1 | NP_001192849.1 | 1 - 2 | 113 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BOLA-DOA| Gene Symbol: BOLA-DOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205920.1 | NP_001192849.1 | 1 - 2 | 113 | 108 | |
Set Membership: |
|