Assay Details
| Gene Symbol: ZNF496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205929.1 | NP_001192858.1 | 1 - 2 | 394 | 75 | |
| GenBank mRNA | FN908756.1 | - | 3 - 4 | 710 | 75 | |
| FN908757.1 | - | 2 - 3 | 497 | 75 | ||
More Information
Set Membership: |
|

ZNF496
Genomic Map