Assay Details
| Gene Symbol: ZNF496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205929.1 | NP_001192858.1 | 6 - 7 | 1009 | 76 | |
| GenBank mRNA | FN908757.1 | - | 7 - 8 | 1112 | 76 | |
More Information
Set Membership: |
|

ZNF496
Genomic Map