Assay Details
| Gene Symbol: INO80D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205940.2 | NP_001192869.2 | 1 - 2 | 218 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

INO80D| Gene Symbol: INO80D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205940.2 | NP_001192869.2 | 1 - 2 | 218 | 61 | |
Set Membership: |
|