Assay Details
| Gene Symbol: ATF7IP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205967.1 | NP_001192896.1 | 8 - 9 | 1561 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATF7IP2| Gene Symbol: ATF7IP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205967.1 | NP_001192896.1 | 8 - 9 | 1561 | 60 | |
Set Membership: |
|