Assay Details
| Gene Symbol: AHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206026.1 | NP_001192955.1 | 8 - 9 | 1062 | 74 | |
| GenBank mRNA | AY143368.1 | - | 8 - 9 | 1024 | 74 | |
More Information
Set Membership: |
|

AHR
Genomic Map