Assay Details
| Gene Symbol: EGLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206046.2 | NP_001192975.2 | 3 - 4 | 1261 | 75 | |
| GenBank mRNA | EE242828.1 | - | 3 - 4 | 708 | 75 | |
More Information
Set Membership: |
|

EGLN1
Genomic Map